Learn More
BINDER™ Coated Heat Exchanger
SDP
Catalog No. 502875691 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO 1600
KB PRO 260
KB PRO 470
KB PRO 720, KB PRO 1060
KBF PRO 130
KBF PRO 1600
KBF PRO 260
KBF PRO 470
KBF PRO 720, KBF PRO 1060
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5691 KB PRO 260
50-287-5745 KB PRO 470
50-287-5746 KB PRO 720, KB PRO 1060
50-287-5749 KBF PRO 130
50-287-5623 KB PRO 1600
50-287-5624 KBF PRO 260
50-287-5722 KBF PRO 470
50-287-5657 KBF PRO 720, KBF PRO 1060
50-287-5690 KBF PRO 1600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 50-287-5691 Supplier BINDER™ Supplier No. 80122462
Add to Cart
Add to Cart
Heat exchanger that is coated in plastic to prevent damage.
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
For Use With (Equipment) KB PRO 260
Material Aluminum
Type Heat Exchanger