Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ELISA Carbonate Coating Buffer

Catalog No. CB01100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB01100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB01100 Supplier Thermo Scientific™ Supplier No. CB01100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.

  • ELISA Carbonate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.
  • The buffer is a 50 mM carbonate buffer, pH 9.4.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Coating Buffer
Quantity 100 mL
Conjugate Enzyme Labeled
Form Concentrate
For Use With (Application) ELISA
Content And Storage Buffer. Store at RT.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.