Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt

Catalog No. 20307625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-307-625G 25 g
20-307-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-307-625G Supplier Sigma Aldrich Supplier No. 20307625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed in the synthesis of a metal-rich layered telluride, TaCo2Te2, which possesses ideal characteristics for photovoltaic applications, for surface scanning studies, and which may give clean and atomically flat surfaces.

Specifications

CAS 7440-48-4
Density 8.9 g/mL (at 25°C)
Boiling Point 2900°C
Molecular Formula Co
Linear Formula Co
Quantity 25 g
MDL Number MFCD00010935
Molecular Weight (g/mol) 58.93
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.