Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt AA Standard, 1000 ppm Co in 3% HNO3, Ricca Chemical
SDP

Catalog No. ACO1KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ACO1KN100 100 mL Natural LDPE Bottle
ACO1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ACO1KN100 Supplier Ricca Chemical Company Supplier No. ACO1KN100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-48-4 , 7697-37-2 , 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Co
CAS Min % 93.0, 4.89, 0.09
CAS Max % 97.0, 5.09, 0.1
Equivalence 1 mL = 1 mg Co
Physical Form Liquid
Color Pink
pH < 2
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Cobalt AA Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.