Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt aluminum oxide, Co 39-42%

Catalog No. p-7049663
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1768622 100 g
AA1768636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1768622 Supplier Thermo Scientific Chemicals Supplier No. 01768622
Only null left
Add to Cart

CAS: 12672-27-4 | Al2Co2O5 | 251.824 g/mol

It is used in the catalytic deposition of nitrous oxide over calcinated cobalt aluminum hydrotalcites. The activity increased with an increase in cobalt concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12672-27-4
Molecular Formula Al2Co2O5
Molecular Weight (g/mol) 251.824
MDL Number MFCD00016016
InChI Key CRHLEZORXKQUEI-UHFFFAOYSA-N
Synonym cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiide
PubChem CID 159384
IUPAC Name dialuminum;cobalt(2+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 176.89
Concentration or Composition (by Analyte or Components) Co 39 to 41%
Odor Odorless
Chemical Name or Material Cobalt aluminum oxide
Hazard Category H317-H319-H334-H341-H350-H360
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P272-P280-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 235-762-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.