Learn More
Description
- Detects the presence of moisture when exposed to humidity of 50% or greater.
- Strip will turn from blue to light pink/white color in the presence of humidity.
- Great for detecting moisture in hard to reach places.
- 100 Strips/Vial, 12 Vials/Pack
Specifications
Specifications
| Color Indicator | Blue to Light Pink/White |
| pH Range | <7 |
| For Use With (Application) | Moisture Detection |
| Parameter Specifications | Detects moisture when exposed to 50% humidity or greater |
| Quantity | 100 Pk., 12 Pk./Cs |
| Type | Cobalt Chloride (CoCl2) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
