Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) carbonate, 99.5% (metals basis)

Catalog No. aa4274518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4274518 50 g
AA4274530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4274518 Supplier Thermo Scientific Chemicals Supplier No. 04274518
Only null left
Add to Cart

CAS: 513-79-1 | CCoO3 | 118.94 g/mol

Cobalt(II) carbonate is a precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and is also used in the manufacture of paints and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-79-1
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00010940
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv
PubChem CID 10565
IUPAC Name cobalt(2+);carbonate
SMILES [Co++].[O-]C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water,alcohol and methyl acetate.
Formula Weight 118.94
Density 4.13 g/mL
Chemical Name or Material Cobalt(II) carbonate
Hazard Category H302-H317-H334-H341-H350i-H360F
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 208-169-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.