Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Standard, 1mL = 1mg Co, 1000ppm Co, Ricca Chemical
SDP

Catalog No. 220016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2200-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2200-16 Supplier Ricca Chemical Company Supplier No. 220016

Chemical Identifiers

CAS 10026-22-9 , 7732-18-5
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1,000ppm Co
Equivalence 1 mL = 1 mg Co
Physical Form Liquid
Color Pink
Quantity 500 mL
Chemical Name or Material Cobalt Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.