Learn More
Ansell Cobalt™ Ultra Nitrile Gloves
Catalog No. 19166010 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-166-010 Small
19-166-011 Medium
19-166-012 Large
19-166-013 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-166-010 Supplier Ansell Supplier No. N171
This item is not returnable. View return policy
Thin construction for enhanced sensitivity and smart packaging with 200 gloves per box
Manufactured using an ultra-soft and stretchy nitrile formulation for excellent durability and comfort. Its thin construction results in enhanced tactile sensitivity, while textured fingers provide a firm grip. Smartly packed 200 gloves per box to save money and reduce environmental waste.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AQL 1.5, FDA, ASTM F 1671, ISO9001 QMS
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 3 mil
Thickness (Metric) 0.07 mm
Tensile Strength 20.5
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.