Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Chloride, Colorimetric Solution (CS), 1 mL = 59.5 mg CoCl2.6H2O, 59-60.0 mg/mL, Spectrum™ Chemical
SDP

Catalog No. 18601789 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-601-789 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-601-789 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C186500ML
Add to Cart
Add to Cart

C-186, 7791-13-1

Spectrum™ Chemical Cobaltous Chloride, Colorimetric Solution (CS), 1 ml = 59.5 mg CoCl2.6H2O is an acidic aqueous solution used in the preparation of the colorimetric standards for certain drugs, and for the carbonization tests with sulfuric acid that are specified in several monographs.

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

CAS Max % 6%, 93%
Quantity 500 mL
UN Number UN1789
Concentration or Composition (by Analyte or Components) 59-60.0 mg/mL
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.