Learn More
LaMotte™ COD Colorimeter Multi-Range Reagent Systems
Measure levels of Chemical Oxygen Demand using the SMART Spectro, the original SMART colorimeter or the SMART 2 colorimeter
$93.46
Specifications
For Use With (Application) | Measure levels of chemical oxygen demand |
---|---|
For Use With (Equipment) | SMART spectro, original SMART or SMART2 colorimeters |
Includes | 25 high grade, optical quality 16mm vials with TFE lined caps |
Type | Water Testing System |
Description
- Each package contains 25 high-grade, optical quality 16mm vials with TFE-lined caps
Specifications
Measure levels of chemical oxygen demand | |
SMART spectro, original SMART or SMART2 colorimeters | |
25 high grade, optical quality 16mm vials with TFE lined caps | |
Water Testing System |