Description
- Adjustable nose cone limits movement
- Helps animal remain comfortable
- Decreases injury to animal and researcher
- Dark black nose cone helps animal feel secure
- Nose protrudes through cone front, tail exits through rear hatch opening
Specifications
Specifications
| Construction | Adjustable black nose cone |
| For Use With (Application) | Observe Animal Behavior During Research |
| Type | Animal Holder |
| For Use With (Equipment) | Rat |
| Size | Medium |