Learn More
Brady™ Code Reader™ 2600 Barcode Readers
Catalog No. 19104472 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
Bluetooth, Battery
Type:
Barcode Reader Palm Kit
Catalog No. Includes Type
19-104-472 Bluetooth, Battery Barcode Reader Palm Kit
1 options
Catalog No. 19-104-472 Supplier Brady™ Supplier No. 133083
Reads all 1D and 2D barcode symbologies
Reads all 1D and 2D barcode symbologies
  • Easy to use, lightweight, and high performing
  • Features durable, rechargeable battery plus programmable buttons for custom workflow
  • Battery status LED indicators with fuel gauge
  • User feedback with vibration, audible tones and LED
  • Patented dual field optics, both high density and wide field in same unit
  • High speed, omnidirectional reading of 1D and 2D barcodes
  • Unequaled performance on rounded, curved and shiny surfaces
  • Multiple programmable buttons for customized work flow processes
  • Easy to clean, disinfectant ready, IP65 housing for healthcare use
  • Powerful data management capability with JavaScript
  • Available in Handle and Palm kits
Specifications
Includes Bluetooth, Battery
Type Barcode Reader Palm Kit