Learn More
Brady™ Code Reader™ 900FD Barcode Readers
Catalog No. 19104467 Shop All Brady Corporation Products
Change view
Click to view available options
Includes:
RS232 Cable, Stand, US Power Supply
USB cable, stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-104-467 RS232 Cable, Stand, US Power Supply
19-104-466 USB cable, stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-467 Supplier Brady™ Supplier No. 133073
Used for data collection and tracking applications
  • High speed, omnidirectional imaging of 1D barcode symbologies
  • Can upgrade to 2D or stacked barcodes with purchase of license key (serial number of scanner required for license purchase)
  • Wide Area Image Sensor
  • Manual or Automatic triggering
  • 2 programmable indicators: LED and tone
  • IP54 Housing
Specifications
For Use With (Application) Basic data entry, tracking, shipping and receiving, UID initiatives, WIP
Includes RS232 Cable, Stand, US Power Supply
Type 900FD Barcode Reader