Learn More
Cayman Chemical Coelenterazine 400a, 70217-82-2, 1 mg
SDP
Supplier:  Cayman Chemical 161571
Encompass_Preferred
A bisdeoxy derivative of coelenterazine that produces an emission maximum of 395 nm following conversion by Renilla luciferase; commonly used in BRET2 protocols to evaluate protein-protein interactions
Catalog No. NC0772401
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.