Learn More
Research Products International Corp Coenzyme A, Free Acid, 10 Milligrams
SDP
Supplier:  Research Products International Corp C708000.01
Encompass_Preferred
Extracted from yeast. Coenzyme A is a coenzyme, notable for its role in the synthesis and oxidation of fatty acids, and the oxidation of pyuvate in the citric acid cycle.
Catalog No. 50-488-624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.