Learn More
AdipoGen CoenzymeQ10
SDP
Supplier:  AdipoGen CDXC0661G001
Encompass_Preferred
Chemical. CAS 303-98-0. Formula C59H90O4. MW 863.34. Synthetic. Coenzyme Q10 CoQ10 is a component of the electron transport chain of the mitochondria and participates in aerobic cellular respiration, generating energy in the form of ATP. In its reduced form, CoQ10 acts as an antioxidant, preventing the formation of reactive oxygen species and displaying neuroprotective activity. CoQ10 deficiencies have been associated with heart failure, hypertension, parkinsonism, mitochondrial encephalomyopathies and other chronic diseases. Shown to modulate the nitric oxide pathway, inhibit LDL oxidation and suppress the down regulation of NOSIII endothelial nitric oxide synthase, eNOS and up-regulation of NOSII inducible nitric oxide synthase, iNOS.
Catalog No. 50-256-1354
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.