Learn More
TARGETMOL CHEMICALS INC COH34 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T108595MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. COH34 (1-[(E)-(4-methylphenyl)sulfanyliminomethyl]naphthalen-2-ol) is a selective inhibitor of PARG with an IC50 of 0.37 nM and a Kd of 0.547 uM. COH34 prolongs PARylation at DNA lesions and traps DNA repair factors. purity: 98%
Catalog No. 50-258-1639