Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL17A1 Recombinant Rabbit Monoclonal Antibody (15G9)
GREENER_CHOICE

Catalog No. FEMA550189 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550189 Supplier Invitrogen™ Supplier No. MA550189
  • $485.00
    $446.65 / Each of 1

    Save $38.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

COL17A1 Recombinant Monoclonal Antibody for ELISA, Western Blot

COL17A1 is the alpha chain of type XVII collagen. Unlike most collagens, collagen XVII is a transmembrane protein, and a structural component of hemidesmosomes, multiprotein complexes at the dermal-epidermal basement membrane zone that mediate adhesion of keratinocytes to the underlying membrane. COL17A1 is a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Moreover, COL17A1 is localized to tissues containing type I collagen so, like other members of this collagen family, it may serve to maintain the integrity of the extracellular matrix. Diseases associated with COL17A1 dysfunction include epithelial recurrent erosion dystrophy and epidermolysis bullosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL17A1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 15G9
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4
Gene COL17A1
Gene Accession No. Q9UMD9
Gene Alias 120 kDa linear IgA dermatosis antigen; 120 kDa linear IgA disease antigen; 120 kDa linear IgA disease antigen homolog; 180 kDa bullous pemphigoid antigen 2; 97 kDa LAD antigen; 97 kDa linear IgA bullous dermatosis antigen; 97 kDa linear IgA disease antigen; alpha 1 type XVII collagen; BA16H23.2; bA16H23.2 (collagen, type XVII, alpha 1 (BP180)); BP180; BPA-2; Bpag; BPAG2; Bullous pemphigoid antigen 2; bullous pemphigoid antigen 2 (180kD); Col17a1; Collagen alpha-1(XVII) chain; collagen type XVII alpha 1; collagen XVII, alpha-1 polypeptide; collagen, type XVII, alpha 1; ERED; LABD97; LAD-1; Linear IgA bullous disease antigen of 97 kDa; Linear IgA disease antigen 1; procollagen, type XVII, alpha 1; type XVII collagen alpha-1
Gene Symbols COL17A1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human COL17A1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1308
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.