Learn More
Invitrogen™ COL19A1 Polyclonal Antibody

Description
COL19A1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | COL19A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | COL19A1 |
| Gene Accession No. | Q0VF58, Q14993 |
| Gene Alias | a1 chain of type XIX collagen; col19a1; COL9A1L; collagen alpha 1 (Y) chain; collagen alpha-1(XIX) chain; Collagen alpha-1(Y) chain; collagen type XIX alpha 1 chain; collagen XIX, alpha-1 polypeptide; collagen, type XIX, alpha 1; D6S228E; procollagen, type XIX, alpha 1 |
| Gene Symbols | COL19A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.