Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL1A2 Mouse anti-Human, Porcine, Clone: 3D5E8, Proteintech
SDP

Catalog No. 501737167 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7167 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7167 Supplier Proteintech Group Inc Supplier No. 667611IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Type I collagen is a member of group I collagen (fibrillar forming collagen).

Specifications

Antigen COL1A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3D5E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene COL1A2
Gene Accession No. P08123
Gene Alias Alpha 2 type I collagen, COL1A2, Collagen alpha 2(I) chain, collagen, type I, alpha 2, OI4
Gene Symbols COL1A2
Host Species Mouse
Immunogen COL1A2 Fusion Protein Ag6281
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100626716, 1278
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.