Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL22A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555200 Supplier Invitrogen™ Supplier No. PA555200
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKEELEEIAS EPKSAHVFHV SDFNAIDKIR GKLRRRLCEN VLCPSVRVEG DRFKHTNGGT KEITGFDLMD LFSVKEILGK RENGAQSSYV RMG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes member of the collagen family which is thought to contribute to the stabilization of myotendinous junctions and strengthen skeletal muscle attachments during contractile activity. It belongs to the fibril-associated collagens with interrupted triple helix (FACIT) subset of the collagen superfamily, which associate with collagen fibers through their C-terminal collagenous domains and mediate protein-protein interactions through their N-terminal noncollagenous domains. The encoded protein is deposited in the basement membrane zone of the myotendinous junction which is present only at the tissue junctions of muscles, tendons, the heart, articular cartilage, and skin. A knockdown of the orthologous zebrafish gene induces a muscular dystrophy by disruption of the myotendinous junction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL22A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL22A1
Gene Accession No. Q8NFW1
Gene Alias 2310067L16Rik; C80743; COL22A1; Collagen alpha-1(XXII) chain; collagen type XXII alpha 1; collagen type XXII alpha 1 chain; collagen, type XXII, alpha 1; procollagen, type XXII, alpha 1
Gene Symbols COL22A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL22A1. Recombinant protein control fragment (Product #RP-93895).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.