Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585634 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585634 Supplier Invitrogen™ Supplier No. PA585634
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (86%), Rat (90%), Bovine (82%). Positive Control: SK-N-SH. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Collagen IV alpha-1 (COL4A1) encodes the major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene COL4A1
Gene Accession No. P02462, P02463
Gene Alias alpha 1 type IV collagen; alpha1(IV) collagen; arresten; Bru; BSVD; COL4A1; Col4a-1; COL4A1 NC1 domain; collagen alpha-1(IV) chain; collagen IV, alpha-1 polypeptide; collagen of basement membrane, alpha-1 chain; collagen type IV alpha 1 chain; collagen, type IV, alpha 1; Del(8)44H; Del(8)Bru44H; procollagen, type IV, alpha 1; RATOR; Raw; retinal anterior wiring; Svc
Gene Symbols COL4A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus of human COL4A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1282, 12826
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.