Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL8A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535077 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535077 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535077 Supplier Invitrogen™ Supplier No. PA535077
  • $616.00
    $489.65 / Each of 1

    Save $126.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

COL8A2 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene encodes the alpha 2 chain of type VIII collagen. The protein is a major component of the basement membrane of the corneal endothelium and forms homo- or heterotrimers with alpha 1 (VIII) type collagens. Defects in this gene are associated with Fuchs endothelial corneal dystrophy and posterior polymorphous corneal dystrophy type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL8A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene COL8A2
Gene Accession No. P25067, P25318
Gene Alias AI429819; COL8A2; Collagen alpha-2(VIII) chain; collagen type VIII alpha 2; collagen VIII, alpha-2 polypeptide; collagen, type VIII, alpha 2; endothelial collagen; FECD; FECD1; PPCD; PPCD2; procollagen type VIII alpha 2; procollagen, type VIII, alpha 2
Gene Symbols COL8A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 571-599 amino acids from the C-terminal region of human COL8A2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1296, 329941
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.