Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs

Catalog No. BP1065A
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
Format:
Bioprocess Container
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
BP1065A 1 L Bioprocess Container
BP1065E 20 L Bioprocess Container
BP1065C 10 L Bioprocess Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. BP1065A Supplier Thermo Scientific™ Supplier No. BP1065A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Minimize prep time and maximize production for media fill trials with this ready-to-use soya broth in BioProcess Containers.

Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Tryptone Soya Broth in BPCs. Simply connect the BioProcess Containers into your existing workflow to maximize process simulations and minimize secondary contamination.

Effective

  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Just plug and play
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labour costs

Easy-to-Use

  • Innovative bag design and connector options for easy process integration
  • Standard, ready-to-use, off-the-shelf product range
  • No mixing, no sterilization, long shelf life at ambient storage
  • No Mycoplasma contamination
  • Qualified microbial recovery performance
  • Comprehensive validation package for media, bag/components and filling process
  • No filter blockages
  • No mixing required
  • No sterilization
  • No WFI (water for injection) required
  • No vessel cleaning

Pharmacopoeial-compliant Tryptone Soya Broth and Animal Derived Component Free (ADCF) vegetable peptone-based formulations are available.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

SmartSite™ is a trademark or registered trademark of the Cardinal Healthcare Inc.

ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear, straw
Content And Storage 2 to 25°C
Description Cold Filterable Tryptone Soya Broth in BPCs
Format Bioprocess Container
Product Type Microbiology Media
Includes Includes the following connectors: LUER lock female +ö and SmartSite, needle-free valve additional sampling port
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.