Learn More
Hach Company Cold finger for 75 mL tube - accessory for BD-40 and BD-46 block digesters
Supplier: Hach Company 18032
Cold finger designed for 75 mL digestion tubes. Recommended accessory for use with BD-40 and BD-46 block digesters. Sold individually.
- Fits 75 mL digestion tubes.
- Compatible with select block digesters.
- Sold individually.
- Replacement accessory for block digestion workflows.