Learn More
CPC™ HFC35 Polysulfone Hose Barb Coupling Bodies
Catalog No. 05113150 Shop All Colder Products Company Products
Change view
Click to view available options
Tubing Size:
1/2 in.
3/8 in.
Description:
Non-valved
Valved
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tubing Size Description
05-113-150 3/8 in. Valved
05-113-151 1/2 in. Valved
05-113-152 1/2 in. Non-valved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-113-150 Supplier CPC™ Supplier No. HFCD17635
Free floating
Specifications
Description Valved
Material Polysulfone
Tubing Size 3/8 in.
Product Type In-line coupling; Free floating; EPDM seal options; Vacuum to 125 psi, 8.6 bar; Valve Spring: 316 Stainless Steel