Learn More
Description
- Large tow handle and wheels
- Molded handles
- Environmentally friendly insulation
Specifications
Specifications
| Capacity (English) | 100 qt. |
| Includes | Large tow molded handle, wheels and Rustproof, leak resistant no-tilt drain |
| For Use With (Application) | For easy movement across all-terrain |
| Product Type | Wheeled Cooler |
| Color | Blue |
| Capacity (Metric) | 94.6 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.