Learn More
Description
- Holds ice up to five days at temperatures up to 90°F
- Channel drain
- Two-way handles
Specifications
Specifications
| Capacity (English) | 62 qt. |
| Includes | Channel drain and two-way handles |
| Product Type | Xtreme Cooler |
| Color | Blue |
| Capacity (Metric) | 58.6 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.