Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Petrifilm™ Coliform Count Plates
SCI_ED

Catalog No. S30684 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
1000 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30684 1000 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30684 Supplier 3M™ Petrifilm™ Supplier No. 6416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accurate, sample-ready plates to detect coliform levels in food, dairy, and environmental samples in just 24 hours.

  • Rapid Results: Detect coliform bacteria in food and environmental samples within 24 hours
  • Sample-Ready Format: No need to prepare media or agar dishes, add sample and incubate
  • High Throughput: Ideal for food processors, contract labs, and quality assurance teams
  • Validated Accuracy:
    • Official Methods of Analysis (OMA): #986.33 (Milk), #989.10 (Dairy Products), #991.14 (Food) NF
    • VALIDATION Certifications by AFNOR: 3M 01/02-09/89 A, B, and C for a wide range of human foods, pet food, and environmental samples
  • Automation-Ready: Compatible with the 3M™ Petrifilm™ Plate Reader for fast, standardized results recording
  • Target Organism: Detects coliform bacteria
  • Application: Raw materials, processed foods, finished products, environmental and equipment testing

Specifications

Detects Coliform
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Identifying coliform levels for food processors and contract labs
For Use With (Equipment) 3M™ Petrifilm™ plate reader
Quantity 1000 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.