Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colistin Sodium Methanesulfonate, G-Biosciences
SCI_ED

Catalog No. S666982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S666982 5 g
S000025 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S666982 Supplier G-Biosciences Supplier No. RC985
Add to Cart
Add to Cart

Colistin is a polymyxin antibiotic effective against gram-negative bacteria. It has been used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination.

Colistin has also been implicated in researching effective treatment for P. aeruginosa infections. Colistin functions by targeting cell membranes and altering their permeability.

Chemical Identifiers

Molecular Weight (g/mol) 1759.90 g/mol

Specifications

Quantity 5 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.