Learn More
Medchemexpress LLC HY-P1061A 1mg Medchemexpress, Colivelin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1061A1MG
Encompass_Preferred
Medchemexpress, HY-P1061A 1mg Colivelin (TFA) CAS: Colivelin (TFA) is a neuroprotective peptide and activator of STAT3 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4150