Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ COLL2-1 (Dog) ELISA Kit

Catalog No. 89032266 Shop All Abnova Corporation Products
Change view
Click to view available options
Target Species:
Canine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Species
89-032-266 Canine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-266 Supplier Abnova™ Supplier No. KA4901
Add to Cart
Add to Cart

Competitive enzyme immunoassay for the quantitative measurement of a degradation fragment containing a 9 amino acid sequence (HRGYPGLDG) from type II collagen molecule.

  • For quantification of Coll2-1, a degradation product of type II collagen, in canine serum
  • For research use only (RUO) - Not for use in diagnostic procedures

Specifications

Calibration Range 31.25 to 2000 nM
Description COLL2-1 (Dog) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of a degradation fragment containing a 9 amino acid sequence (HRGYPGLDG) from type II collagen molecule.
Conjugate HRP
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Serum
Target Species Canine
For Use With (Application) Quantification
Gene ID (Entrez) 1280
Kit Contents PLT Coll2-1 pre-coated Microtiter plate 96 wells, STD Coll2-1 standard, 2000 nM calibrator, CTL1 Positive control, 1000 nM, AB1 Coll2-1 polyclonal antibody Conc. (100x), AB2 Goat anti-rabbit HRP antibody Conc. (100x), DIL1 Diluent for STD samples and AB1, DIL2 Diluent for AB2, WASH Washing buffer Conc. (10x), TMP Substrate solution, STOP - Stop solution
Regulatory Status For research use only (RUO)
Sample Volume 50 μL (Prediluted)
Storage Requirements Store at 4°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.