Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Collagen (Calf)

Catalog No. 89940148 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-148 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-940-148 Supplier Abnova™ Supplier No. P5216

For use as substrate

#REF!

Specifications

Concentration 6mg/mL
For Use With (Application) Enzyme Assay Substrate
Formulation In 75mM sodiuim citrate, pH 3.6 - 4.0 and 0.01% thimerisol
Name Collagen (Calf)
Quantity 10 mL
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name COLLAGEN
Assays Platelet aggregation assays?
Biological Activity Less than 20 minutes gel time.
Species Bovine
Protein Tag Untagged
Expression System Native protein purified from Calf Skin
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.