Learn More
Rockland Immunochemicals COLLAGEN I ALPHA 1 PROPEPTIDE
SDP
Supplier:  Rockland Immunochemicals 600401D19
Encompass
Collagen I alpha 1 propeptide Antibody 100ul.Host:Rabbit.Target Species:Human.Known Cross Reactivity:Human, mouse, rat.Clonality:Polyclonal.
Catalog No. NC1773627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.