Learn More
Description
Native mouse collagen I/III
Source:Murine tail tendons
Amino Acid Sequence:UniProt P08121, P11087, Q01149
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | ELISA |
| Formulation | Essentially salt free |
| Molecular Weight (g/mol) | 300 kDa |
| Quantity | 0.01 g |
| Storage Requirements | Store at 4°C. After reconstitution store at -20°C. |
| Gene Alias | Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1 |
| Gene Symbol | COL1A1 |
| Reconstitution | Use 0.05-0.5 M acetic acid, pH 2.5 at 4°C. Dissolved collagen retains immunologic properties of native collagen types I and III. Thermal denaturation converts dissolved collagen to gelatin. |
| Buffer | Essentially salt free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
