Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Collagen I, Native, Bovine, FITC Conjugated, 5 mg

Catalog No. 502703804
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-3804 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-3804 Supplier Abcam Supplier No. AB3142015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

FITC-conjugated full length native bovine collagen protein.

  • Protein Type: Native
  • Species Reactivity: Bovine
  • Form: Lyophilized
  • Storage Buffer: 100% Tris buffered saline
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Collagen I
Substrate Properties Protein-based Substrate
Shipping Condition Blue Ice
Label or Dye FITC
Buffer Tris buffered saline
Concentration 5 mg/mL
Content And Storage Store at -20°C.
Form Lyophilized
Detection Method Fluorescence
Formulation Lyophilized from 100% Tris buffered saline
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.