Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Collagen I, Native Bovine, Full Length, FITC, 1 mg

Catalog No. 502703805
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-3805 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-3805 Supplier Abcam Supplier No. AB3142011MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-conjugated native bovine collagen protein for research applications.

  • Protein Type: Native
  • Source: Cow (Bovine)
  • Conjugation: FITC
  • Form: Lyophilized; Constituents: 100% Tris buffered saline
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Collagen I (Bovine)
Label or Dye FITC
Shipping Condition Blue Ice
Form Lyophilized
Detection Method Fluorescence
Concentration 5 mg/mL
Buffer Tris buffered saline
Content And Storage Store at -20°C.
Formulation Lyophilized from 100% Tris buffered saline
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.