Learn More
Description
Native rat collagen I
Source:Rat
Amino Acid Sequence:UniProt P02466, P02454
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | ELISA |
| Formulation | Lyophilised from aqueous 0.1 M acetic acid |
| Quantity | 0.01 g |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Reconstitution | Reconstitute in ice cold 0.5 M acetic acid, pH 2.5. Solubility - 10 mg/mL. |
| Buffer | Lyophilised from aqueous 0.1 M acetic acid |
| Product Line | Novus |
| Purity or Quality Grade | >90%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
