Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Collagen I Native Protein
SDP

Catalog No. NBP197266 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197266 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197266 Supplier R&D Systems™ Supplier No. NBP197266

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Western Blot, ELISA, Immunohistochemistry

Native COL1A1. Source: Human Placenta (Accession #: P02452) The Collagen I alpha 1 Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Specifications

Accession Number P02452, P02452, P02452, P02452
Concentration 1.0 mg/mL
For Use With (Application) ELISA, Immunohistochemistry, Western Blot
Formulation 0.5 M Acetic Acid
Gene ID (Entrez) 1277
Name Collagen I alpha 1 protein
Quantity 0.5 mg
Source Human serum
Storage Requirements Store at 4 C. Do not freeze.
Gene Symbol COL1A1
Cross Reactivity Human
Research Category Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells
Product Line Novus
Purity or Quality Grade Multi-step
Protein Collagen I alpha 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.