Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. p-4543271 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS306P1ABX 100 μL Concentrated 1mg/mL
MS306P 1 mL Concentrated 200μg/mL
MS306R7 7 mL Prediluted -
MS306P0 100 μL Concentrated 200μg/mL
MS306PABX 200 μL Concentrated 1mg/mL
MS306P1 500 μL Concentrated 200μg/mL
MS306R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS306P1ABX Supplier Epredia Supplier No. MS306P1ABX
Only null left
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Collagen II Ab-3, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 6B3

Isotype: IgG1

Species Reactivity: Human, Mouse, Chicken and Salamander. Others not known.

Epitope: Triple helix of type II collagen

Immunogen: Purified type II collagen; obtained from the adult chicken sternum by limited pepsinization

Positive Control: Cartilage in lung or fetus

Cellular Localization: Cartilage matrix

In Western blotting, Ab-3 reacts with the TCA fragment of lathyritic type II collagen after digestion with mammalian collagenase. It also reacts with pepsin-digested type II collagen. Immunoblotting of CNBr peptides of collagen II shows that MAb 6B3 reacts with CB 11 fragment which is the site of immunogenic and arthritogenic epitopes along the intact type II molecule. In Western blotting (but not ELISA), Ab-3 recognizes alpha1(II) as well as alpha3(XI) chain both of which have identical primary structure.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Collagen II Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6B3
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified type II collagen; obtained from the adult chicken sternum by limited pepsinization
Quantity 100 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Salamander
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.