Learn More
Abcam Collagen III, Native Human, Full Length, Unconjugated, 500 μg
Description
- Target Molecule: Collagen III (COL3A1)
- Source: Native Human
- Predicted Molecular Weight: 138 kDa
- Tested Applications: ELISA, IP, SDS-PAGE, WB
- Form: Solution
- Concentration: 1 mg/mL
- Storage: -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Collagen III (COL3A1) |
| Format | Vial |
| Form | Solution |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Human |
| Quantity | 500 μg |
| For Use With (Application) | ELISA, IP, SDS-PAGE, Western blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.