Learn More
Abcam Collagen III Peptide, Synthetic, Human, >70% Purity, 1 mg
Description
- Species Reactivity: Human
- Tested Applications: BL
- Purity: >70%
- Form: Liquid
- Conjugation: Unconjugated
- Constituents: 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Collagen III (Human) |
| Reactivity | Human |
| Label or Dye | Unconjugated |
| Kit Contents | 1 mg Human Collagen III peptide |
| Product Type | Blocker and Diluent |
| Form | Liquid |
| Detection Location | In-Blot Detection |
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate ethoxylated, pH6.75 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.