Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A1 Monoclonal Antibody (3G3)
GREENER_CHOICE

Catalog No. PIMA536227 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536227 Supplier Invitrogen™ Supplier No. MA536227
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293T whole cell, human Hela whole cell, human A549 whole cell. IHC: human lung cancer tissue, human placenta tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Collagen IV alpha-1 (COL4A1) encodes the major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3G3
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene COL4A1
Gene Accession No. P02462
Gene Alias alpha 1 type IV collagen; alpha1(IV) collagen; arresten; Bru; BSVD; COL4A1; Col4a-1; COL4A1 NC1 domain; collagen alpha-1(IV) chain; collagen IV, alpha-1 polypeptide; collagen of basement membrane, alpha-1 chain; collagen type IV alpha 1 chain; collagen, type IV, alpha 1; Del(8)44H; Del(8)Bru44H; procollagen, type IV, alpha 1; RATOR; Raw; retinal anterior wiring; Svc
Gene Symbols COL4A1
Host Species Mouse
Immunogen E.coli-derived human Collagen IV recombinant protein (Position: G1445-T1669).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1282
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.