Learn More
Advanced Biomatrix Human Collagen Powder, Type III, Non-sterile, 10 mg
Supplier:  Advanced Biomatrix 501910MG
Encompass_Preferred
Type III Collagen provides structure and strength to connective tissue, is found many places in the body, especially skin, lung, intestinal walls and the walls of blood vessels. This Type III Collagen product is isolated from human placenta and is purified using a multi-step process with approximately 85% Type III collagen with the remainder being comprised of Type I collagen. The product is supplied as a non-sterile powder. A Certificate of Analysis is available with the purchase of each product. Type III collagen product is provided in a user-friendly packaging for use and storage.Type III Collagen is typically used as a thin coating on tissue culture surfaces or as a control. Specific instructions are found in the Directions for Use. This product is generally used in vitro as a substrate scaffold to enhance cell attachment, adherence and proliferation. Type III collagen may be used to culture a variety of cell types.
Catalog No. 50-196-0272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.