Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collagen V Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102416 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102416 Supplier Invitrogen™ Supplier No. PA5102416
  • $634.00
    $478.65 / Each of 1

    Save $155.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Collagen V alpha 1.

COL5A1 encodes an alpha chain for one of the lowabundance fibrillar collagens. Fibrillar collagen molecules aretrimers that can be composed of one or more types of alpha chains.Type V collagen is found in tissues containing type I collagen andappears to regulate the assembly of heterotypic fibers composed ofboth type I and type V collagen. COL5A1 product is closelyrelated to type XI collagen and it is possible that the collagenchains of types V and XI constitute a single collagen type withtissue-specific chain combinations. Mutations in this gene areassociated with Ehlers-Danlos syndrome, types I and II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen V
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution
Western Blot (WB)  1:500-1:2,000
Immunohistochemistry (Paraffin) (IHC (P))  1:50-1:200
Immunocytochemistry (ICC/IF)  1:100-1:500
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COL5A1
Gene Accession No. O88207, P20908, Q9JI03
Gene Alias AB collagen; AI413331; alpha-1 type V collagen; alpha1(V) collagen; COL5; Col5a1; Collagen 5; collagen alpha 1 (V); collagen alpha-1(V) chain; collagen type 5 alpha 1; collagen type V alpha 1; collagen type V alpha 1 chain; collagen, type V, alpha 1; EDSC; pro-alpha1(V) collagen; procollagen alpha 1(V); procollagen type V alpha 1; procollagen, type V, alpha 1; RP11-263F14.1; Type V Collagen
Gene Symbols COL5A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human COL5A1(Accession P20908), corresponding to amino acid residues E305-L355.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12831, 1289, 85490
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.