Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Collagen V Protein, Native, Human, Full Length, 500 ug

Catalog No. 502709826
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-9826 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-9826 Supplier Abcam Supplier No. AB7537500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Full-length native human collagen V protein suitable for use in IP, WB, ELISA, and SDS-PAGE applications.

  • Protein Type: Native
  • Source: Human
  • Full Length: Yes
  • Applications: ELISA, IP, SDS-PAGE, WB
  • Concentration: 2.7 mg/mL

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Collagen V
Substrate Properties Protein-based Substrate
Concentration 2.7 mg/mL
Content And Storage Store at -20°C.
Form Solution
Label or Dye Unconjugated
Cross Reactivity Human
Quantity 500 μg
For Use With (Application) IP, Western Blot, ELISA, SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.