Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Collagen VI alpha 1 Native Protein
SDP

Catalog No. NBP197270 Shop All R&D Systems Products
Change view
Click to view available options
0.5mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP197270 0.5mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197270 Supplier Novus Biologicals™ Supplier No. NBP197270
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native COL6A1. Source: Human Placenta (Accession #: P12109) The Collagen VI alpha 1 Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Specifications

Accession Number P12109
Formulation 0.5M Acetic Acid
Gene ID (Entrez) 1291
Name Collagen VI protein
Source Human serum
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.