Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Collagenase (C. histolyticum)

Catalog No. 89940150 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
4000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-150 4000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-940-150 Supplier Abnova™ Supplier No. P5295
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Collagenase (C. histolyticum)
Preparation Method Native protein chromatographically purified from Clostridium histolyticum
Quantity 4000 U
Storage Requirements Store at 4°C on dry atmosphere. For primary cell isolation, culture and tissue dissociation: Reconstituted in balanced salt solutions (such as Hank's, Earle's and others) at concentrations from 0.05% to 0.5% (w/v). For enzymetic assay: Reconstituted in 0.05 M TES buffer with 0.36 mM calcium chloride, pH 7.5 at a concentration of 1.0 mg/ml.
Regulatory Status RUO
Common Name COLLAGENASE
Assays Collagenase Assay; Pancreatic and parotid acini isolation and collagen structural analysis.
Biological Activity ≥500 units per mg dry weight. One Unit releases one micromole of L-leucine equivalents from collagen in 5 hours at 37°C, pH 7.5.
Species Clostridium species
Protein Tag None
Expression System Native protein chromatographically purified from Clostridium histolyticum
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.