Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Collagenase Inhibitor Assay Kit, Fluorometric, 100 Tests

Catalog No. 502588470
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-8470 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-8470 Supplier Abcam Supplier No. AB211108100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay kit designed for measuring MMP1 inhibitor activity.

  • Detection method: Fluorescent
  • Applications: Enzyme activity assay
  • Sample types: Inhibitor compounds
  • Results type: Semi-Quantitative
  • Citations: Cited in > 13 publications
  • Compatible equipment: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Collagenase
No. of Reactions 100 Tests
For Use With (Equipment) Microplate Reader
Format Kit
Detection Method Fluorescence
Sample Type Inhibitor compounds
Quantity 100 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.